Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017401.2 | NP_059097.2 | 3 | 715 | |
GenBank mRNA | AF176098.1 | - | 3 | 488 | |
AJ251804.2 | - | 3 | 459 | ||
AK137747.1 | - | 2 | 1666 | ||
BC056213.1 | - | 3 | 507 | 30024844 | |
BC056996.1 | - | 1 | 2248 |