Gene Transcripts

Gene Symbol
Cts8
Entrez Gene ID
56094
Gene Name
cathepsin 8
Gene Aliases
CTS2, Epcs68, Epcs70
Chromosome Location
Chr. 13: 61246747 - 61255348 on Build GRCm38
UniGene ID
Mm.342861
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019541.3 NP_062414.3 5 612
GenBank mRNA AF250840.1 - 5 624
AK049474.1 - 5 613
AY014780.1 - 5 611
BC068241.1 - 5 602 30019120

View Full Product Details