Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019995.3 | NP_064379.1 | 6 | 407 | |
GenBank mRNA | AF139179.1 | - | 6 | 411 | |
AK050619.1 | - | 7 | 431 | ||
AK083379.1 | - | 7 | 431 | ||
AK188434.1 | - | 4 | 211 | ||
AK210270.1 | - | 3 | 191 | ||
AK212629.1 | - | 5 | 264 | ||
BC052702.1 | - | 6 | 396 | 6400922 |