Gene Transcripts

Gene Symbol
Prl5a1
Entrez Gene ID
28078
Gene Name
prolactin family 5, subfamily a, member 1
Gene Aliases
1600013P04Rik, D13Wsu14e, PLP-L, Prlpl
Chromosome Location
Chr. 13: 28142484 - 28151595 on Build GRCm38
UniGene ID
Mm.155058
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023746.4 NP_076235.1 2 225
GenBank mRNA AF226611.1 - 2 172
AK005447.1 - 2 226
BC051672.1 - 2 212 30017843

View Full Product Details