Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282444.1 | NP_001269373.1 | 5 | 1070 | |
NM_001282445.1 | NP_001269374.1 | 4 | 748 | ||
NM_006795.3 | NP_006786.2 | 3 | 927 | ||
XM_011544739.1 | XP_011543041.1 | 4 | 745 | ||
GenBank mRNA | AF001434.1 | - | 3 | 878 | |
AF099011.1 | - | 869 | |||
AK097842.1 | - | 5 | 1035 | ||
AK124613.1 | - | 2 | 1534 | ||
AK297654.1 | - | 4 | 606 | ||
AK299096.1 | - | 4 | 659 | ||
AK300326.1 | - | 4 | 708 | ||
AK302930.1 | - | 4 | 700 | ||
AK312316.1 | - | 3 | 780 | ||
AY007161.1 | - | 3 | 674 | ||
BC104799.1 | - | 3 | 819 | 8143802 | |
BC104825.1 | - | 3 | 819 | 8143828 |