Gene Transcripts

Gene Symbol
Slco1a4
Entrez Gene ID
28250
Gene Name
solute carrier organic anion transporter family, member 1a4
Gene Aliases
AI785519, Oatp1a4, Oatp2, Slc21a5
Chromosome Location
Chr. 6: 141805438 - 141856283 on Build GRCm38
UniGene ID
Mm.255586
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030687.1 NP_109612.1 17 2684
GenBank mRNA AB031814.2 - 17 2684
AK050401.1 - 1 389

View Full Product Details