Gene Transcripts

Gene Symbol
Nrep
Entrez Gene ID
27528
Gene Name
neuronal regeneration related protein
Gene Aliases
AI325076, D0H4S114, Harp, P311, PTZ17, SEZ17
Chromosome Location
Chr. 18: 33437019 - 33464029 on Build GRCm38
UniGene ID
Mm.407415
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109988.1 NP_001103458.1 5 1713
NM_001109989.2 NP_001103459.1 5 1636
NM_001267717.1 NP_001254646.1 5 1625
NM_053078.4 NP_444308.1 4 1751
GenBank mRNA AK048361.1 - 4 1551
AK133769.1 - 3 2845
BC054762.1 - 4 1533 6414312
D45203.1 - 1 1148
X70398.1 - 4 1549

View Full Product Details