Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145705.3 | NP_663751.2 | 5 | 727 | |
GenBank mRNA | AF214013.1 | - | 5 | 718 | |
AF518764.1 | - | 5 | 692 | ||
AK016456.1 | - | 5 | 660 | ||
AK028952.1 | - | 5 | 698 | ||
AK090324.1 | - | 5 | 694 | ||
AK144592.1 | - | 5 | 656 | ||
AK169480.1 | - | 3 | 242 | ||
BC017142.1 | - | 2 | 1002 | ||
BC030347.1 | - | 5 | 653 | 5003804 |