Gene Transcripts

Gene Symbol
Ift88
Entrez Gene ID
21821
Gene Name
intraflagellar transport 88
Gene Aliases
AW552028, Tg737, Tg737Rpw, TgN737Rpw, Ttc10, flexo, fxo, orpk, polaris
Chromosome Location
Chr. 14: 57423657 - 57517936 on Build GRCm38
UniGene ID
Mm.4653
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009376.2 NP_033402.2 15 1398
GenBank mRNA AK135110.1 - 15 1393
AK162011.1 - 2 103
AK168395.1 - 14 1307
BC012250.1 - 15 1378 4019334
L31959.1 - 15 1343

View Full Product Details