Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009410.2 | NP_033436.1 | 5, 6 | 758 | |
GenBank mRNA | AB006074.1 | - | 5, 6 | 759 | |
AK029987.1 | - | 2, 3 | 151 | ||
AK037640.1 | - | 5, 6 | 659 | ||
AK164443.1 | - | 5, 6 | 659 | ||
AK170291.1 | - | 6, 7 | 565 | ||
AK171286.1 | - | 6, 7 | 565 | ||
BC099689.1 | - | 5, 6 | 573 | 30731611 |