Gene Transcripts

Gene Symbol
Kdm6a
Entrez Gene ID
22289
Gene Name
lysine (K)-specific demethylase 6A
Gene Aliases
Utx
Chromosome Location
Chr. X: 18162533 - 18279791 on Build GRCm38
UniGene ID
Mm.257498
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009483.1 NP_033509.1 2, 3 578
GenBank mRNA AJ002730.1 - 1, 2 37
AK080370.1 - 2, 3 597
AK144654.1 - 2, 3 604
AK164941.1 - 2, 3 610
BC053433.1 - 2, 3 578 6516368

View Full Product Details