Gene Transcripts

Gene Symbol
Zfp93
Entrez Gene ID
22755
Gene Name
zinc finger protein 93
Gene Aliases
Znf235
Chromosome Location
Chr. 7: 24270418 - 24277794 on Build GRCm38
UniGene ID
Mm.459431
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009567.4 NP_033593.1 4 2790
GenBank mRNA AK028905.1 - 4 2627
AK040325.1 - 4 2721
AK132229.1 - 4 2791

View Full Product Details