Gene Transcripts

Gene Symbol
Cln8
Entrez Gene ID
26889
Gene Name
ceroid-lipofuscinosis, neuronal 8
Gene Aliases
mnd
Chromosome Location
Chr. 8: 14881335 - 14901720 on Build GRCm38
UniGene ID
Mm.254027
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012000.3 NP_036130.1 3 2595
GenBank mRNA AF125308.1 - 3 2085
AK049145.1 - 3 2268
AK089692.1 - 4 2036
AK128890.1 - 3 2262
AK164268.1 - 3 2268
AK166905.1 - 3 3020
AK171446.1 - 3 2277
BC021625.1 - 3 2097 4188527

View Full Product Details