Gene Transcripts

Gene Symbol
Vsnl1
Entrez Gene ID
26950
Gene Name
visinin-like 1
Gene Aliases
VILIP, Vnsl1
Chromosome Location
Chr. 12: 11325245 - 11436649 on Build GRCm38
UniGene ID
Mm.27005
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012038.4 NP_036168.1 4 1739
GenBank mRNA AJ489331.1 - 4 1694
AK005091.2 - 3 1300
AK144454.1 - 4 1670
AY101375.1 - 3 1440

View Full Product Details