Gene Transcripts

Gene Symbol
Rbck1
Entrez Gene ID
24105
Gene Name
RanBP-type and C3HC4-type zinc finger containing 1
Gene Aliases
AL033326, HOIL-1, HOIL-1L, UIP28, Ubce7ip3
Chromosome Location
Chr. 2: 152316334 - 152332643 on Build GRCm38
UniGene ID
Mm.182145
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083921.1 NP_001077390.1 12 2296
NM_019705.3 NP_062679.2 13 2137
GenBank mRNA AF124663.1 - 11 1906
AK085559.1 - 12 2286
AK088591.1 - 12 2259
AK146176.1 - 13 2311
AK166075.1 - 12 2264
AK210683.1 - 18
BC032089.1 - 5 1741
BC034555.1 - 13 2133 4192310

View Full Product Details