Gene Transcripts

Gene Symbol
Morf4l1
Entrez Gene ID
21761
Gene Name
mortality factor 4 like 1
Gene Aliases
MORFRG15, MRG15, TEG-189, Tex189, mKIAA4002
Chromosome Location
Chr. 9: 90091669 - 90114820 on Build GRCm38
UniGene ID
Mm.426209
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039147.2 NP_001034236.1 11 1088
NM_024431.3 NP_077751.1 10 971
GenBank mRNA AB042241.1 - 10 820
AF319620.1 - 10 819
AF319621.1 - 11 936
AK010528.1 - 10 739
AK016414.1 - 10 938
AK041833.1 - 2 1282
AK079115.1 - 6 421
AK146210.1 - 10 806
AK146785.1 - 9 701
AK150925.1 - 10 772
AK153545.1 - 5 404
AK167079.1 - 10 730
AK168327.1 - 10 806
AK168585.1 - 10 751
AK168608.1 - 10 753
AK168674.1 - 10 802
AK169065.1 - 10 803
AK184930.1 - 4 312
AK205535.1 - 4 312
AK213016.1 - 4 312
AK214961.1 - 4 313
AK216220.1 - 4 312
AK216852.1 - 3 313
AK220441.1 - 8 3747
BC003894.1 - 10 823 3593982
BC017619.1 - 10 823 4923923
BC083094.1 - 10 757
BC083118.1 - 10 832 5371036
BC085103.1 - 11 938 6490291
BC092293.1 - 10 835 5068516
BC103783.1 - 10 835 6529024

View Full Product Details