Gene Transcripts

Gene Symbol
Pex3
Entrez Gene ID
83519
Gene Name
peroxisomal biogenesis factor 3
Gene Aliases
Chromosome Location
Chr. 1: 7685209 - 7726238 on Build Rnor_6.0
UniGene ID
Rn.11773
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031350.1 NP_112640.1 6 580
GenBank mRNA AB035306.1 - 6 580
BC062046.1 - 6 671 5597999

View Full Product Details