Gene Transcripts
- Gene Symbol
- Wap
- Entrez Gene ID
- 114596
- Gene Name
- whey acidic protein
- Gene Aliases
- Chromosome Location
- Chr. 14: 86897948 - 86900412 on Build Rnor_6.0
- UniGene ID
- Rn.9978
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053751.4 |
NP_446203.1
|
4 |
489 |
|
GenBank mRNA |
J00802.1 |
- |
4 |
489 |
|
View Full Product Details