Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138870.1 | NP_620225.1 | 1 | 204 | |
XM_006228036.2 | XP_006228098.1 | 2 | 231 | ||
XM_006228037.2 | XP_006228099.1 | 2 | 231 | ||
GenBank mRNA | AB029495.1 | - | 1 | 204 |