Gene Transcripts

Gene Symbol
Sfxn5
Entrez Gene ID
261737
Gene Name
sideroflexin 5
Gene Aliases
BBG-TCC
Chromosome Location
Chr. 4: 116992645 - 117112141 on Build Rnor_6.0
UniGene ID
Rn.161851
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153298.1 NP_695210.1 3, 4 300
XM_008763051.1 XP_008761273.1 3, 4 893
GenBank mRNA AB056724.1 - 3, 4 317

View Full Product Details