Gene Transcripts

Gene Symbol
Nudt1
Entrez Gene ID
117260
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 1
Gene Aliases
Mth1
Chromosome Location
Chr. 12: 16391578 - 16398746 on Build Rnor_6.0
UniGene ID
Rn.10669
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057120.1 NP_476461.1 2 259
XM_006248911.2 XP_006248973.1 3 310
GenBank mRNA D49977.1 - 2 259

View Full Product Details