Gene Transcripts
- Gene Symbol
- LYPLA2
- Entrez Gene ID
- 11313
- Gene Name
- lysophospholipase II
- Gene Aliases
- APT-2, APT2, DJ886K2.4
- Chromosome Location
- Chr. 1: 23790789 - 23795539 on Build GRCh38
- UniGene ID
- Hs.533479
- Species
- Homo sapiens
- Gene Symbol
- LYPLA2P2
- Entrez Gene ID
- 388499
- Gene Name
- lysophospholipase II pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 19: 7878619 - 7880232 on Build GRCh38
- UniGene ID
- Hs.730360
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033250.1 |
|
1 |
418 |
|
View Full Product Details