Gene Transcripts

Gene Symbol
AUP1
Entrez Gene ID
550
Gene Name
ancient ubiquitous protein 1
Gene Aliases
Chromosome Location
Chr. 2: 74526648 - 74529897 on Build GRCh38
UniGene ID
Hs.411480
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181575.4 NP_853553.1 3 545
NR_126510.1 3 545
NR_126511.1 3 545
XM_005264392.3 XP_005264449.1 2 490
GenBank mRNA AF100746.1 - 3 318
AF100753.1 - 3 345
AF100754.1 - 3 345
AF165515.1 - 3 325
AK023983.1 - 3 323
AK055566.1 - 2 444
AK293121.1 - 3 364
AK301398.1 - 3 344
AK304886.1 - 2 487
AK308231.1 - 3 346
AK310159.1 - 3 346
BC001658.1 - 3 408 3050204
BC033646.2 - 3 360
CR749661.1 - 2 416
CR999537.1 - 3 531
CX164419.1 - 3 327

View Full Product Details