Gene Transcripts

Gene Symbol
Cpe
Entrez Gene ID
25669
Gene Name
carboxypeptidase E
Gene Aliases
CARBE, Cph
Chromosome Location
Chr. 16: 26906716 - 27014811 on Build Rnor_6.0
UniGene ID
Rn.7149
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013128.1 NP_037260.1 6 1163
GenBank mRNA BC085762.1 - 6 1162
J04625.1 - 6 1118
M31602.1 - 6 1134
X51406.1 - 6 1163

View Full Product Details