Gene Transcripts

Gene Symbol
Pitpna
Entrez Gene ID
29525
Gene Name
phosphatidylinositol transfer protein, alpha
Gene Aliases
Pitpn
Chromosome Location
Chr. 10: 63731767 - 63772049 on Build Rnor_6.0
UniGene ID
Rn.9771
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017231.1 NP_058927.1 7, 8 643
GenBank mRNA BC070945.1 - 7, 8 671
FQ235021.1 - 1, 2 44
M25758.1 - 7, 8 643

View Full Product Details