Gene Transcripts

Gene Symbol
Aqp7
Entrez Gene ID
29171
Gene Name
aquaporin 7
Gene Aliases
Chromosome Location
Chr. 5: 57358327 - 57372239 on Build Rnor_6.0
UniGene ID
Rn.11111
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019157.2 NP_062030.2 3 418
GenBank mRNA AB000507.2 - 3 418
AY120935.1 - 2 157
AY157737.1 - 2 157
BC072695.1 - 3 378 7104684

View Full Product Details