Gene Transcripts

Gene Symbol
LSM5
Entrez Gene ID
23658
Gene Name
LSM5 homolog, U6 small nuclear RNA and mRNA degradation associated
Gene Aliases
YER146W
Chromosome Location
Chr. 7: 32485333 - 32495258 on Build GRCh38
UniGene ID
Hs.424908
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130710.1 NP_001124182.1 5 247
NM_001139499.1 NP_001132971.1 5 486
NM_012322.2 NP_036454.1 5 310
NR_024466.1 5 1057
GenBank mRNA AF182291.1 - 5 271
AJ238097.1 - 5 264
AK022772.1 - 5 284
AK024217.1 - 5 293
BC005938.1 - 5 270 4132686
BF700164.1 - 5 245
DA736378.1 - 5 477

View Full Product Details