Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012434.4 | NP_036566.1 | 3 | 461 | |
XM_005248710.3 | XP_005248767.1 | 3 | 483 | ||
XM_005248711.2 | XP_005248768.1 | 3 | 715 | ||
XM_011535750.2 | XP_011534052.1 | 3 | 419 | ||
XM_017010730.1 | XP_016866219.1 | 3 | 715 | ||
XM_017010731.1 | XP_016866220.1 | 3 | 565 | ||
GenBank mRNA | AF244577.1 | - | 4 | 718 | |
AJ387747.1 | - | 3 | 603 | ||
AK075320.1 | - | 3 | 415 | ||
AK294068.1 | - | 3 | 420 | ||
AK315789.1 | - | 3 | 419 | ||
BC020961.2 | - | 3 | 420 | 3847279 | |
BP288187.1 | - | 3 | 461 |