Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031784.2 | NP_113972.2 | 3 | 576 | |
XM_006232991.2 | XP_006233053.1 | 3 | 829 | ||
XM_006232992.2 | XP_006233054.1 | 3 | 1911 | ||
XM_006232993.2 | XP_006233055.1 | 3 | 718 | ||
XM_008761337.1 | XP_008759559.1 | 3 | 580 | ||
XM_008761338.1 | XP_008759560.1 | 3 | 829 | ||
GenBank mRNA | AF032872.2 | - | 3 | 576 |