Gene Transcripts

Gene Symbol
CFAP20
Entrez Gene ID
29105
Gene Name
cilia and flagella associated protein 20
Gene Aliases
C16orf80, EVORF, GTL3, fSAP23
Chromosome Location
Chr. 16: 58113588 - 58129425 on Build GRCh38
UniGene ID
Hs.532755
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013242.2 NP_037374.1 6 1174
NR_136632.1 6 1342
GenBank mRNA AF093680.1 - 6 1153
AK027099.1 - 6 1097
BC005152.2 - 6 1143 3528301

View Full Product Details