Gene Transcripts

Gene Symbol
NDUFA1
Entrez Gene ID
4694
Gene Name
NADH:ubiquinone oxidoreductase subunit A1
Gene Aliases
CI-MWFE, MWFE, ZNF183
Chromosome Location
Chr. X: 119871771 - 119876666 on Build GRCh38
UniGene ID
Hs.534168
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004541.3 NP_004532.1 3 363
GenBank mRNA BC000266.1 - 3 260
BQ680674.1 - 3 264
U54993.1 - 3 255
X81900.1 - 3 239

View Full Product Details