Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001190799.1 | NP_001177728.1 | 13 | 2839 | |
NM_001190801.1 | NP_001177730.1 | 14 | 2758 | ||
NM_014264.4 | NP_055079.3 | 14 | 2935 | ||
XM_005262701.2 | XP_005262758.1 | 14 | 2795 | ||
XM_017007662.1 | XP_016863151.1 | 14 | 3094 | ||
XM_017007663.1 | XP_016863152.1 | 14 | 2993 | ||
GenBank mRNA | AB006972.1 | - | 14 | 2849 | |
AK302858.1 | - | 14 | 2758 | ||
AK303399.1 | - | 13 | 2773 | ||
AK314238.1 | - | 14 | 2903 | ||
BC036023.1 | - | 14 | 2903 | 5273226 | |
Y13115.1 | - | 14 | 2801 |