Gene Transcripts

Gene Symbol
PDIA5
Entrez Gene ID
10954
Gene Name
protein disulfide isomerase family A member 5
Gene Aliases
PDIR
Chromosome Location
Chr. 3: 123067009 - 123162106 on Build GRCh38
UniGene ID
Hs.477352
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006810.3 NP_006801.1 2 275
NR_028444.1 2 275
GenBank mRNA BC001625.1 - 2 222
BM549365.1 - 2 275
D49490.1 - 2 175

View Full Product Details