Gene Transcripts

Gene Symbol
RAB25
Entrez Gene ID
57111
Gene Name
RAB25, member RAS oncogene family
Gene Aliases
CATX-8, RAB11C
Chromosome Location
Chr. 1: 156061149 - 156070514 on Build GRCh38
UniGene ID
Hs.632469
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020387.3 NP_065120.2 5 969
NR_133653.1 4 773
GenBank mRNA AF083124.1 - 5 927
AF274025.1 - 5 948
BC004416.1 - 3 459
BC009831.2 - 5 943 3926839
BC033322.1 - 5 933 4775725

View Full Product Details