Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318934.1 | NP_001305863.1 | 9 | 1231 | |
NM_001318935.1 | NP_001305864.1 | 9 | 1231 | ||
NM_003071.3 | NP_003062.2 | 9 | 1231 | ||
NM_139048.2 | NP_620636.1 | 9 | 1231 | ||
XM_011513091.1 | XP_011511393.1 | 9 | 1231 | ||
XM_011513092.1 | XP_011511394.1 | 9 | 1231 | ||
XM_011513093.1 | XP_011511395.1 | 9 | 1231 | ||
XM_017007078.1 | XP_016862567.1 | 9 | 1210 | ||
XM_017007079.1 | XP_016862568.1 | 9 | 1231 | ||
GenBank mRNA | AB209052.1 | - | 9 | 1013 | |
AK291231.1 | - | 9 | 1178 | ||
BC005260.1 | - | 9 | 1195 | ||
BC015498.1 | - | 9 | 1187 | ||
BC030976.1 | - | 9 | 1231 | ||
BC044659.1 | - | 9 | 1199 | 6015181 | |
L34673.1 | - | 9 | 1190 |