Gene Transcripts

Gene Symbol
OR9Q1
Entrez Gene ID
219956
Gene Name
olfactory receptor family 9 subfamily Q member 1
Gene Aliases
Chromosome Location
Chr. 11: 58023881 - 58181587 on Build GRCh38
UniGene ID
Hs.689841
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005212.3 NP_001005212.1 3 492
GenBank mRNA BC110819.1 - 3 492

View Full Product Details