Gene Transcripts

Gene Symbol
OR4D9
Entrez Gene ID
390199
Gene Name
olfactory receptor family 4 subfamily D member 9
Gene Aliases
OR11-253
Chromosome Location
Chr. 11: 59514913 - 59515857 on Build GRCh38
UniGene ID
Hs.553757
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004711.1 NP_001004711.1 1 918

View Full Product Details