Gene Transcripts

Gene Symbol
OR4N2
Entrez Gene ID
390429
Gene Name
olfactory receptor family 4 subfamily N member 2
Gene Aliases
OR14-13, OR14-8
Chromosome Location
Chr. 14: 19803770 - 19828372 on Build GRCh38
UniGene ID
Hs.512490
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004723.2 NP_001004723.1 2 808

View Full Product Details