Gene Transcripts

Gene Symbol
PLEKHG6
Entrez Gene ID
55200
Gene Name
pleckstrin homology and RhoGEF domain containing G6
Gene Aliases
MyoGEF
Chromosome Location
Chr. 12: 6310436 - 6328506 on Build GRCh38
UniGene ID
Hs.631660
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144856.1 NP_001138328.1 14 1783
NM_001144857.1 NP_001138329.1 13 1951
NM_018173.3 NP_060643.2 14 1701
XM_005253704.4 XP_005253761.1 14 1798
XM_006718985.3 XP_006719048.1 14 1756
XM_011520967.2 XP_011519269.1 14 1673
XM_017019555.1 XP_016875044.1 14 1798
GenBank mRNA AF289613.1 - 2 243
AK001527.1 - 14 1702
AK023741.1 - 14 1783
AK095373.1 - 13 1951
BC104751.1 - 14 1637 40035012
BC105961.1 - 14 1637 40035014
BC109095.2 - 14 1637 40034695

View Full Product Details