Gene Transcripts

Gene Symbol
AIRE
Entrez Gene ID
326
Gene Name
autoimmune regulator
Gene Aliases
AIRE1, APECED, APS1, APSI, PGA1
Chromosome Location
Chr. 21: 44285838 - 44298219 on Build GRCh38
UniGene ID
Hs.129829
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000383.3 NP_000374.1 14 1973
GenBank mRNA AB006682.1 - 14 1931
AB006683.1 - 8 1449
AB006685.1 - 9 1367
Z97990.1 - 14 1924

View Full Product Details