Gene Transcripts

Gene Symbol
OR10T2
Entrez Gene ID
128360
Gene Name
olfactory receptor family 10 subfamily T member 2
Gene Aliases
OR1-3
Chromosome Location
Chr. 1: 158398522 - 158399466 on Build GRCh38
UniGene ID
Hs.553588
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004475.1 NP_001004475.1 1 490

View Full Product Details