Gene Transcripts

Gene Symbol
OR13J1
Entrez Gene ID
392309
Gene Name
olfactory receptor family 13 subfamily J member 1
Gene Aliases
OR9-2
Chromosome Location
Chr. 9: 35869463 - 35870401 on Build GRCh38
UniGene ID
Hs.527480
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004487.1 NP_001004487.1 1 22
GenBank mRNA BC136715.1 - 1 85
BC136716.1 - 1 85 9020706

View Full Product Details