Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001816.3 | NP_001807.2 | 2 | 215 | |
XM_011526340.2 | XP_011524642.1 | 2 | 215 | ||
XM_011526341.1 | XP_011524643.1 | 2 | 215 | ||
XM_011526342.1 | XP_011524644.1 | 2 | 215 | ||
XM_017026194.1 | XP_016881683.1 | 2 | 215 | ||
XM_017026195.1 | XP_016881684.1 | 2 | 215 | ||
XM_017026196.1 | XP_016881685.1 | 2 | 215 | ||
XM_017026198.1 | XP_016881687.1 | 2 | 215 | ||
GenBank mRNA | AK297012.1 | - | 2 | 183 | |
AK312652.1 | - | 2 | 212 | ||
BC026263.1 | - | 2 | 183 | 4618311 | |
D90064.1 | - | 2 | 199 | ||
DA464901.1 | - | 2 | 215 | ||
M33326.1 | - | 2 | 199 | ||
X52378.1 | - | 2 | 145 |