Gene Transcripts

Gene Symbol
SAMD1
Entrez Gene ID
90378
Gene Name
sterile alpha motif domain containing 1
Gene Aliases
Chromosome Location
Chr. 19: 14088417 - 14090420 on Build GRCh38
UniGene ID
Hs.140309
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138352.1 NP_612361.1 1406
GenBank mRNA AY453840.1 - 1406
BC007384.2 - 4 500
BC030129.2 - 4 679
BC065477.1 - 4 647
BC080588.1 - 4 500

View Full Product Details