Gene Transcripts
- Gene Symbol
- LCNL1
- Entrez Gene ID
- 401562
- Gene Name
- lipocalin like 1
- Gene Aliases
- Chromosome Location
- Chr. 9: 136982993 - 136985758 on Build GRCh38
- UniGene ID
- Hs.695210
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK097466.1 |
- |
1 |
358 |
|
AK127236.1 |
- |
1 |
2456 |
|
BC042845.1 |
- |
1 |
1036 |
|
View Full Product Details