Gene Transcripts

Gene Symbol
Rpl9
Entrez Gene ID
29257
Gene Name
ribosomal protein L9
Gene Aliases
Chromosome Location
Chr. 14: 44524419 - 44527613 on Build Rnor_6.0
UniGene ID
Rn.107914
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007598.3 NP_001007599.3 5, 6 423
GenBank mRNA BC060589.1 - 5, 6 427
BC086561.1 - 4, 5 405 7304371
FQ210154.1 - 5, 6 423
FQ211371.1 - 5, 6 423
FQ212163.1 - 5, 6 422
FQ212174.1 - 5, 6 423
FQ212384.1 - 5, 6 427
FQ212431.1 - 5, 6 422
FQ216865.1 - 5, 6 423
FQ220825.1 - 5, 6 421
FQ221003.1 - 5, 6 423
FQ221028.1 - 5, 6 423
FQ221603.1 - 5, 6 423
FQ221794.1 - 5, 6 425
FQ221999.1 - 5, 6 425
FQ222130.1 - 5, 6 425
FQ222146.1 - 5, 6 423
FQ222204.1 - 5, 6 423
FQ222278.1 - 5, 6 421
FQ222508.1 - 5, 6 425
FQ222509.1 - 5, 6 423
FQ222609.1 - 5, 6 425
FQ222630.1 - 5, 6 422
FQ222743.1 - 5, 6 423
FQ223008.1 - 5, 6 437
FQ223059.1 - 5, 6 423
FQ223395.1 - 5, 6 425
FQ223420.1 - 5, 6 421
FQ223651.1 - 5, 6 418
FQ224555.1 - 5, 6 423
FQ228532.1 - 5, 6 435
FQ229128.1 - 5, 6 421
X51706.1 - 4, 5 405

Gene Symbol
LOC100360449
Entrez Gene ID
100360449
Gene Name
ribosomal protein L9-like
Gene Aliases
Chromosome Location
Chr. 3: 4282206 - 4282911 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002729147.2 XP_002729193.1 1 429

Gene Symbol
LOC100364457
Entrez Gene ID
100364457
Gene Name
ribosomal protein L9-like
Gene Aliases
Chromosome Location
Chr. 1: 77829695 - 77830412 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003748774.3 XP_003748822.1 1 436

View Full Product Details