Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001007637.1 | NP_001007638.1 | 6 | 701 | |
XM_006232604.2 | XP_006232666.1 | 6 | 993 | ||
XM_006232605.2 | XP_006232667.1 | 6 | 996 | ||
XM_006232607.2 | XP_006232669.1 | 6 | 883 | ||
XM_006232608.2 | XP_006232670.1 | 6 | 913 | ||
GenBank mRNA | BC082018.1 | - | 6 | 701 |