Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001007656.1 | NP_001007657.1 | 8 | 1680 | |
XM_006239796.2 | XP_006239858.1 | 7 | 1563 | ||
XM_008764512.1 | XP_008762734.1 | 7 | 1730 | ||
XM_008764513.1 | XP_008762735.1 | 7 | 1685 | ||
GenBank mRNA | BC083589.1 | - | 8 | 1680 |