Gene Transcripts

Gene Symbol
Mettl23
Entrez Gene ID
287918
Gene Name
methyltransferase like 23
Gene Aliases
RGD1306284
Chromosome Location
Chr. 10: 105787935 - 105793304 on Build Rnor_6.0
UniGene ID
Rn.99722
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008283.1 NP_001008284.1 5 774
XM_006247798.1 XP_006247860.1 5 760
GenBank mRNA BC086594.1 - 5 774

View Full Product Details