Gene Transcripts

Gene Symbol
Slc10a4
Entrez Gene ID
305309
Gene Name
solute carrier family 10, member 4
Gene Aliases
Chromosome Location
Chr. 14: 37764839 - 37770059 on Build Rnor_6.0
UniGene ID
Rn.79638
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008555.1 NP_001008555.1 3 865
GenBank mRNA AY704415.1 - 3 865
AY825923.1 - 3 865

View Full Product Details